• USA Home
  • Search Results

  • benzeneseleninicacidanhydride360131769712011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benzeneseleninic acid anhydride

1 Products
Image of Benzeneseleninic acid anhydride
CAS Number:
Linear Formula:
 C6H5SeOOSeOC6H5
Molecular Weight:
 360.13
 Beilstein/REAXYS Number:
 2332406
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More