• USA Home
  • Search Results

  • benzenesolution78117143211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benzene solution

2 Products
CAS Number:
Empirical Formula (Hill Notation):
 C6H6
Molecular Weight:
 78.11
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Anal. Standard Form
  Special Grade
  Titertest
Show More