• USA Home
  • Search Results

  • benzenesulfonicacidmonohydrate176192615800911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benzenesulfonic acid monohydrate

1 Products
Image of Benzenesulfonic acid monohydrate
CAS Number:
Linear Formula:
 C6H5SO3H · H2O
Molecular Weight:
 176.19
 Beilstein Registry Number:
 4552655
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More