• USA Home
  • Search Results

  • benzoapyrene252315032811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benzo[a]pyrene

4 Products
Synonym:   3,4-Benzopyrene, 3,4-Benzpyrene, Benzo[def]chrysene
Image of Benzo[<I>a</I>]pyrene
CAS Number:
Empirical Formula (Hill Notation):
 C20H12
Molecular Weight:
 252.31
 Beilstein/REAXYS Number:
 1911333
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  Physical Form
  Special Grade
Show More