• USA Home
  • Search Results

  • benzoapyrenesolution252315032811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benzo[a]pyrene solution

4 Products
Image of Benzo[<I>a</I>]pyrene solution
CAS Number:
Empirical Formula (Hill Notation):
 C20H12
Molecular Weight:
 252.31
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Feature
  Purity Grade
  Package quantity
  Anal. Standard Form
  Special Grade
  Concentration
Show More