• USA Home
  • Search Results

  • benzobfluoranthene2523120599211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benzo[b]fluoranthene

3 Products
Synonym:   2,3-Benzfluoranthene, 3,4-Benzofluoranthene, 3,4-Benz[e]acephenanthrylene, 3,4-Benzfluoranthene, Benz[e]acephenanthrylene, Benzo[e]fluoranthene, NSC 89265
Image of Benzo[<I>b</I>]fluoranthene
CAS Number:
Empirical Formula (Hill Notation):
 C20H12
Molecular Weight:
 252.31
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Package quantity
  Anal. Standard Form
  Physical Form
  Special Grade
Show More