• USA Home
  • Search Results

  • benzocaine165199409711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benzocaine

4 Products
Synonym:   4-Aminobenzoic acid ethyl ester, Ethyl 4-aminobenzoate
Image of Benzocaine
CAS Number:
Empirical Formula (Hill Notation):
 C9H11NO2
Molecular Weight:
 165.19
 Beilstein/REAXYS Number:
 638434
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Anal. Standard Form
  Application(s)
  Manufacturer designation
  Physical Form
  Special Grade
Show More