• USA Home
  • Search Results

  • benzofuran32hone13413716934811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benzofuran-3(2H)-one

1 Products
Synonym:   3-Coumaranone
Image of Benzofuran-3(2<I>H</I>)-one
CAS Number:
Empirical Formula (Hill Notation):
 C8H6O2
Molecular Weight:
 134.13
 Beilstein/REAXYS Number:
 115296
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More