• USA Home
  • Search Results

  • benzoicacidsolution122126585011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benzoic acid solution

2 Products
Image of Benzoic acid solution
CAS Number:
Empirical Formula (Hill Notation):
 C7H6O2
Molecular Weight:
 122.12
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Special Grade
  Application(s)
  Purity Grade
Show More