• USA Home
  • Search Results

  • benzoinmethylether22627352462711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benzoin methyl ether

2 Products
Synonym:   α-Methoxy-α-phenylacetophenone, 2-Methoxy-2-phenylacetophenone
Image of Benzoin methyl ether
CAS Number:
Linear Formula:
 C6H5CH(OCH3)COC6H5
Molecular Weight:
 226.27
 Beilstein/REAXYS Number:
 2052112
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More