• USA Home
  • Search Results

  • benzonase12345902565411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benzonase®

1 Products
Synonym:   Endonuclease from Serratia marcescens
Image of Benzonase<SUP>®</SUP>
CAS Number:
 Enzyme Commission (EC) Number:
 3.1.30.2  ( BRENDA | IUBMB )
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More