• USA Home
  • Search Results

  • benzophenone1822211961911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benzophenone

9 Products
Synonym:   Diphenyl ketone, Diphenylmethanone, NSC 8077
Image of Benzophenone
CAS Number:
Linear Formula:
 (C6H5)2CO
Molecular Weight:
 182.22
 Beilstein/REAXYS Number:
 1238185
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Agency/Method
  Potency
  Manufacturer designation
  Melting Point
  Anal. Standard Form
  Application(s)
  Assay
  Physical Form
Show More