• USA Home
  • Search Results

  • benzotriazol1yloxytripyrrolidinophosphoniumhexafluorophosphate5203912862552511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(Benzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate

2 Products
Synonym:   PyBOP®
Image of (Benzotriazol-<WBR>1-<WBR>yloxy)<WBR>tripyrrolidinophosphonium hexafluorophosphate
CAS Number:
Linear Formula:
 C18H28N6OP · PF6
Molecular Weight:
 520.39
 Beilstein/REAXYS Number:
 3584612
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Melting Point
Show More