• USA Home
  • Search Results

  • benzotriazol1yloxytrisdimethylaminophosphoniumhexafluorophosphate442285660233611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate

1 Products
Synonym:   BOP Reagent, BOP, Castros reagent
Image of (Benzotriazol-<WBR>1-<WBR>yloxy)<WBR>tris(dimethylamino)<WBR>phosphonium hexafluorophosphate
CAS Number:
Empirical Formula (Hill Notation):
 C12H22F6N6OP2
Molecular Weight:
 442.28
 Beilstein/REAXYS Number:
 4287025
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More