• USA Home
  • Search Results

  • benzotriazole119129514711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benzotriazole

2 Products
Synonym:   1,2,3-Benzotriazole, 1H-Benzotriazole
Image of Benzotriazole
CAS Number:
Empirical Formula (Hill Notation):
 C6H5N3
Molecular Weight:
 119.12
 Beilstein/REAXYS Number:
 112133
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Application(s)
  Product Line
Show More