• USA Home
  • Search Results

  • benzylacetate1501714011411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benzyl acetate

6 Products
Synonym:   Acetic acid benzyl ester
Image of Benzyl acetate
CAS Number:
Linear Formula:
 CH3COOCH2C6H5
Molecular Weight:
 150.17
 Beilstein/REAXYS Number:
 1908121
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Document Links
  Shelf Life
  Allergen
  Organoleptic property
  Application(s)
  Agency/Method
  Assay
Show More