• USA Home
  • Search Results

  • benzylalcohol1081410051611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benzyl alcohol

21 Products
Synonym:   Benzenemethanol, BnOH
Image of Benzyl alcohol
CAS Number:
Linear Formula:
 C6H5CH2OH
Molecular Weight:
 108.14
 Beilstein/REAXYS Number:
 878307
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
 eCl@ss:
 39023110
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Restriction
  Document Links
  Allergen
  Organoleptic property
  Quality clarifier
  Miscellaneous Specifications
  Potency
  Manufacturer designation
  Biological Source
  Refractive Index
  Anal. Standard Form
  Suitability
  Purity Grade
  Ice Shipping
  Autoignition temp
  Vapor pressure
  Transition temperature
  Shelf Life
  Package quantity
  Explosion limit
  Application(s)
  Solubility
  Product Line
  Agency/Method
  Special Grade
  Assay
Show More

Product #
Description
305197 Benzyl alcohol, anhydrous, 99.8%,
PHR1019 Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material,
402834 Benzyl alcohol, ACS reagent, ≥99.0%,
1061901 Benzyl alcohol, United States Pharmacopeia (USP) Reference Standard,
1.00987 Benzyl alcohol, special grade (benzaldehyde ≤ 0.05%) EMPROVE® EXPERT Ph Eur,BP,JP,NF,
108006 Benzyl alcohol, ReagentPlus®, ≥99%,
08421 Benzyl alcohol, analytical standard,
24122 Benzyl alcohol, puriss., meets analytical specification of Ph. Eur., BP, NF, 99-100.5% (GC),
13160 Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC),
W213713 Benzyl alcohol, natural, ≥98%, FG,
1.02695 Benzyl alcohol, for headspace gas chromatography SupraSolv®,
W213705 Benzyl alcohol, ≥99%, FCC, FG,
8.22259 Benzyl alcohol, for synthesis,
1.37120 Benzyl alcohol, EMPROVE® EXPERT, Ph. Eur., ChP, JP, NF, New
1.09626 Benzyl alcohol, for analysis EMSURE®,
Y0000167 Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard,
ARK2163 Benzyl alcohol, PharmaGrade, USP/NF, Ph. Eur., Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.,
39971 Benzyl alcohol, certified reference material, TraceCERT®,
1.00981 Benzyl alcohol, Emprove® Essential, Ph. Eur., BP, JP, NF,
BX0483 Benzyl alcohol, Meets ACS Specifications GR ACS,
PHL89164 Benzyl alcohol, phyproof® Reference Substance,