• USA Home
  • Search Results

  • benzylbenzoate2122412051411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benzyl benzoate

11 Products
Synonym:   Benzoic acid benzyl ester
Image of Benzyl benzoate
CAS Number:
Linear Formula:
 C6H5COOCH2C6H5
Molecular Weight:
 212.24
 Beilstein/REAXYS Number:
 2049280
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Restriction
  Document Links
  Allergen
  Organoleptic property
  Vapor pressure
  Transition temperature
  Melting Point
  Density
  Autoignition temp
  Special Grade
  Manufacturer designation
  Anal. Standard Form
  Potency
  Application(s)
  Solubility
  Agency/Method
  Assay
  Physical Form
Show More