• USA Home
  • Search Results

  • benzylbutylphthalate312368568711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benzyl butyl phthalate

4 Products
Image of Benzyl butyl phthalate
CAS Number:
Linear Formula:
 2-[CH3(CH2)3O2C]C6H4CO2CH2C6H5
Molecular Weight:
 312.36
 Beilstein/REAXYS Number:
 2062204
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Shelf Life
  Anal. Standard Form
  Physical Form
  Special Grade
Show More