• USA Home
  • Search Results

  • benzylcyanide1171514029411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benzyl cyanide

3 Products
Synonym:   Phenylacetonitrile, α-Tolunitrile
Image of Benzyl cyanide
CAS Number:
Empirical Formula (Hill Notation):
 C8H7N
Molecular Weight:
 117.15
 EC Index Number
 205-410-5
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More