• USA Home
  • Search Results

  • benzyldimethyltetradecylammoniumchloridedihydrate4040714722881711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benzyldimethyltetradecylammonium chloride dihydrate

1 Products
Image of Benzyldimethyltetradecylammonium chloride dihydrate
CAS Number:
Linear Formula:
 CH3(CH2)13N(Cl)(CH3)2CH2C6H5 · 2H2O
Molecular Weight:
 404.07
 Beilstein/REAXYS Number:
 4062599
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More