• USA Home
  • Search Results

  • benzylether1982610350411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benzyl ether

2 Products
Synonym:   Dibenzyl ether
Image of Benzyl ether
CAS Number:
Linear Formula:
 (C6H5CH2)2O
Molecular Weight:
 198.26
 Beilstein/REAXYS Number:
 1911156
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Assay
  Special Grade
  Agency/Method
Show More