• USA Home
  • Search Results

  • benzylpropionate1642012263411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benzyl propionate

3 Products
Image of Benzyl propionate
CAS Number:
Linear Formula:
 C2H5CO2CH2C6H5
Molecular Weight:
 164.20
 Council of Europe no.:
 413
 EC Number:
 FEMA Number:
 2150
 Flavis number:
 9.132
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Potency
  Biological Source
  Document Links
  Allergen
  Organoleptic property
  Special Grade
  Assay
  Agency/Method
Show More