• USA Home
  • Search Results

  • benzyltrimethylammoniumhydroxidesolution1672510085611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Benzyltrimethylammonium hydroxide solution

2 Products
Image of Benzyltrimethylammonium hydroxide solution
CAS Number:
Linear Formula:
 C6H5CH2N(OH)(CH3)3
Molecular Weight:
 167.25
 Beilstein/REAXYS Number:
 3917256
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Density
Show More