• USA Home
  • Search Results

  • beryllium901744041711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Beryllium

76 Products
Synonym:   BE000200, Glucinium
Image of Beryllium
CAS Number:
Empirical Formula (Hill Notation):
 Be
Molecular Weight:
 9.01
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Composite Dimension
  Weight
  Reaction Suitability
  Particle Dimension
  Assay
  Manufacturer designation
  Physical Form
Show More

Product #
Description
GF83666818 Beryllium, foil, thickness 0.01 mm, size 25 × 25 mm, purity 99.8+%,
GF93173676 Beryllium, flake, 10 mm, weight 10 g, purity 99.5%,
GF61658501 Beryllium, foil, thickness 0.5 mm, size 25 × 25 mm, purity 99.8+%,
GF63668010 Beryllium, foil, thickness 0.5 mm, diameter 25 mm, purity 99%,
GF64512097 Beryllium, foil, thickness 2.0 mm, diameter 50 mm, purity 99%,
GF69913525 Beryllium, foil, thickness 1.0 mm, size 25 × 25 mm, purity 99%,
GF70156672 Beryllium, foil, thickness 0.125 mm, size 25 × 25 mm, purity 99.8+%,
GF70622096 Beryllium, foil, thickness 0.125 mm, size 25 × 25 mm, purity 99%,
GF70640838 Beryllium, foil, thickness 0.00025 mm, size 25 × 25 mm, purity 99.9%,
GF72795381 Beryllium, foil, thickness 0.00025 mm, size 12.5 × 12.5 mm, purity 99.9%,
GF76959938 Beryllium, foil, thickness 1.0 mm, diameter 10 mm, purity 99%,
GF79034968 Beryllium, foil, thickness 0.5 mm, size 25 × 25 mm, purity 99%,
GF79370343 Beryllium, foil, thickness 0.5 mm, purity 99%, size 50 × 50 mm,
GF82464161 Beryllium, foil, thickness 0.25 mm, diameter 20 mm, purity 99.8%,
GF84138914 Beryllium, foil, thickness 0.025 mm, size 25 × 25 mm, purity 99.8+%,
GF89047738 Beryllium, foil, thickness 0.025 mm, diameter 15 mm, tolerance 0.2,
GF92548503 Beryllium, foil, thickness 2.0 mm, size 10 × 10 mm, purity 99%,
GF93503178 Beryllium, foil, diameter 25 mm, thickness 0.25 mm, purity 99%,
GF93989389 Beryllium, foil, thickness 0.015 mm, size 25 × 25 mm, purity 99.8+%,
GF96166648 Beryllium, foil, thickness 0.25 mm, diameter 15 mm, purity 99.8%,
GF98781496 Beryllium, foil, thickness 1.0 mm, size 100 × 100 mm, purity 99%,
GF08146481 Beryllium, wire, 1.2 mm diameter, length 0.1 m, purity 99.7%,
GF08665195 Beryllium, foil, thickness 0.25 mm, 29 mm diameter, purity 99.8%,
GF15867917 Beryllium, foil, thickness 0.015 mm, size 10 × 10 mm, purity 99.8+%,
GF18617643 Beryllium, foil, thickness 0.01 mm, size 12.5 × 12.5 mm, purity 99.8+%,
GF21293265 Beryllium, foil, thickness 0.5 mm, size 100 × 100 mm, purity 99%,
GF22066747 Beryllium, foil, thickness 2.0 mm, size 25 × 25 mm, purity 99%,
GF27309562 Beryllium, flake, 10 mm, weight 100 g, purity 99.5%,
GF27759246 Beryllium, wire, 1.2 mm diameter, length 0.2 m, purity 99.7%,
GF30251618 Beryllium, foil, thickness 0.125 mm, 25 mm diameter, purity 99.8%,
GF36694581 Beryllium, foil, thickness 0.25 mm, size 50 × 50 mm, purity 99%,
GF37244538 Beryllium, wire, 1.2 mm diameter, length 0.05 m, purity 99.7%,
GF38618477 Beryllium, flake, 10 mm, weight 50 g, purity 99.5%,
GF38895496 Beryllium, foil, thickness 0.025 mm, 25.4 mm diameter, tolerance 0.2,
GF39190259 Beryllium, foil, thickness 1.0 mm, size 50 × 50 mm, purity 99%,
GF39967644 Beryllium, foil, thickness 0.005 mm, size 12.5 × 12.5 mm, purity 99%,
GF47686304 Beryllium, foil, thickness 1.0 mm, size 25 × 25 mm, purity 99.8+%,
GF50889550 Beryllium, foil, thickness 0.125 mm, 20 mm diameter, purity 99%,
GF52293551 Beryllium, foil, thickness 0.25 mm, size 25 × 25 mm, purity 99%,
GF52842225 Beryllium, foil, thickness 0.25 mm, size 25 × 25 mm, purity 99.8+%,
GF56237418 Beryllium, foil, thickness 0.5 mm, 10 mm diameter, purity 99.8%,
GF56350795 Beryllium, foil, thickness 0.125 mm, 18.8 mm diameter, purity 99.8%,
GF58343182 Beryllium, foil, thickness 2.0 mm, size 50 × 50 mm, purity 99%,
GF67966385 Beryllium, wire reel, 0.3m, diameter 0.356mm, annealed and clean, 99.7%,
GF11051558 Beryllium, wire reel, 0.5m, diameter 0.05mm, annealed and clean, 99.7%,
GF34823224 Beryllium, wire reel, 0.5m, diameter 0.125mm, annealed and clean, 99.7%,
GF24172703 Beryllium, wire reel, 0.5m, diameter 0.25mm, annealed and clean, 99.7%,
GF95797692 Beryllium, wire reel, 0.5m, diameter 0.356mm, annealed and clean, 99.7%,
GF81444764 Beryllium, wire reel, 1m, diameter 0.05mm, annealed and clean, 99.7%,
GF52902840 Beryllium, wire reel, 1m, diameter 0.125mm, annealed and clean, 99.7%,
GF21588565 Beryllium, wire reel, 1m, diameter 0.25mm, annealed and clean, 99.7%,
GF80595793 Beryllium, wire reel, 5m, diameter 0.05mm, annealed and clean, 99.7%,
GF02814702 Beryllium, mesh, 5x5mm, nominal aperture 0.13mm, thickness 0.025mm, wire diameter 0.1mm, 100 wires/inch, open area 50%, electro-formed mesh,
GF12366660 Beryllium, rod, 100mm, diameter 7.0mm, 99%,
GF06987224 Beryllium, wire reel, 0.05m, diameter 0.25mm, annealed and clean, 99.7%,
GF21271025 Beryllium, wire reel, 0.05m, diameter 0.356mm, annealed and clean, 99.7%,
GF28341840 Beryllium, wire reel, 0.05m, diameter 0.5mm, annealed and clean, 99%,
GF65661363 Beryllium, wire reel, 0.05m, diameter 0.6mm, annealed and clean, 99%,
GF03032413 Beryllium, wire reel, 0.05m, diameter 0.75mm, annealed and clean, 99%,
GF41430787 Beryllium, wire reel, 0.05m, diameter 0.85mm, annealed and clean, 99%,
GF53688415 Beryllium, wire reel, 0.1m, diameter 0.05mm, annealed and clean, 99.7%,
GF09275823 Beryllium, wire reel, 0.1m, diameter 0.125mm, annealed and clean, 99.7%,
GF73403142 Beryllium, wire reel, 0.1m, diameter 0.25mm, annealed and clean, 99.7%,
GF58877948 Beryllium, wire reel, 0.1m, diameter 0.356mm, annealed and clean, 99.7%,
GF45831236 Beryllium, wire reel, 0.1m, diameter 0.5mm, annealed and clean, 99%,
GF96781214 Beryllium, wire reel, 0.1m, diameter 0.6mm, annealed and clean, 99%,
GF53985370 Beryllium, wire reel, 0.1m, diameter 0.75mm, annealed and clean, 99%,
GF60949840 Beryllium, wire reel, 0.1m, diameter 0.85mm, annealed and clean, 99%,
GF59478586 Beryllium, wire reel, 0.2m, diameter 0.125mm, annealed and clean, 99.7%,
GF75393388 Beryllium, wire reel, 0.2m, diameter 0.25mm, annealed and clean, 99.7%,
GF59743738 Beryllium, wire reel, 0.2m, diameter 0.356mm, annealed and clean, 99.7%,
GF29665086 Beryllium, wire reel, 0.2m, diameter 0.5mm, annealed and clean, 99%,
GF98865129 Beryllium, wire reel, 0.2m, diameter 0.6mm, annealed and clean, 99%,
GF46045627 Beryllium, wire reel, 0.2m, diameter 0.75mm, annealed and clean, 99%,
GF79324959 Beryllium, wire reel, 0.2m, diameter 0.85mm, annealed and clean, 99%,
265063 Beryllium, chunks, 99.9%,