• USA Home
  • Search Results

  • berylliumsulfatetetrahydrate17714778756611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Beryllium sulfate tetrahydrate

2 Products
Image of Beryllium sulfate tetrahydrate
CAS Number:
Linear Formula:
 BeSO4 · 4H2O
Molecular Weight:
 177.14
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More