• USA Home
  • Search Results

  • bes213251019118111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

BES

5 Products
Synonym:   N,N-Bis(2-hydroxyethyl)-2-aminoethanesulfonic acid, N,N-Bis(2-hydroxyethyl)taurine
Image of BES
CAS Number:
Empirical Formula (Hill Notation):
 C6H15NO5S
Molecular Weight:
 213.25
 Beilstein/REAXYS Number:
 1781572
 EC Number:
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  UV-Absorption Values
  Product Line
  Special Grade
  Solubility
Show More