• USA Home
  • Search Results

  • bestradiol272385028211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

β-Estradiol

4 Products
Synonym:   1,3,5-Estratriene-3,17β-diol, 17β-Estradiol, 3,17β-Dihydroxy-1,3,5(10)-estratriene, Dihydrofolliculin
Image of β-Estradiol
CAS Number:
Empirical Formula (Hill Notation):
 C18H24O2
Molecular Weight:
 272.38
 Beilstein/REAXYS Number:
 1914275
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More