• USA Home
  • Search Results

  • bestradiol6one6ocarboxymethyloxime359423504847611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

β-Estradiol-6-one 6-(O-carboxymethyloxime)

1 Products
Synonym:   β-Estradiol-6-(O-carboxymethyl)oxime, 1,3,5(10)-Estratriene-3,17β-diol-6-one6-(O-carboxymethyloxime), 3,17β-Dihydroxy-1,3,5(10)-estratriene 6-(O-carboxymethyl)oxime, 6-Ketoestradiol 6-(O-carboxymethyloxime)
Image of β-Estradiol-6-one 6-<WBR>(O-<WBR>carboxymethyloxime)
CAS Number:
Empirical Formula (Hill Notation):
 C20H25NO5
Molecular Weight:
 359.42
 Beilstein/REAXYS Number:
 2484202
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More