• USA Home
  • Search Results

  • betacyclodextrin113498758539911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Beta Cyclodextrin

2 Products
Synonym:   β-Cyclodextrin, Caraway, Cycloheptaamylose, Cyclomaltoheptaose, Schardinger β-Dextrin
Image of Beta Cyclodextrin
CAS Number:
Empirical Formula (Hill Notation):
 C42H70O35
Molecular Weight:
 1134.98
 Beilstein/REAXYS Number:
 78623
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Analyte Suitability
Show More