• USA Home
  • Search Results

  • betaglucan12345901272011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Beta Glucan

1 Products
Synonym:   Glucan from baker's yeast (S. cerevisiae)
CAS Number:
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More