• USA Home
  • Search Results

  • betainesolution1171510743711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Betaine solution

1 Products
Image of Betaine solution
CAS Number:
Linear Formula:
 (CH3)3N+CH2COO
Molecular Weight:
 117.15
 Beilstein/REAXYS Number:
 3537113
 MDL number:
 PubChem Substance ID:
Popular Documents:  Bulletin (PDF)  |  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
Show More