• USA Home
  • Search Results

  • betamethasonevalerate47658215244511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Betamethasone valerate

3 Products
Synonym:   1,4-Pregnadiene-11β,17α,21-triol-9α-fluoro-16β-methyl-3,20-dione 17-valerate, 9α-Fluoro-16β-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione 17-valerate, 9α-Fluoro-16β-methylprednisolone 17-valerate, Betamethasone 17-valerate, Betnovate
Image of Betamethasone valerate
CAS Number:
Empirical Formula (Hill Notation):
 C27H37FO6
Molecular Weight:
 476.58
 Beilstein/REAXYS Number:
 4240001
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
  Manufacturer designation
Show More