• USA Home
  • Search Results

  • betasitosterol414718346511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Beta-sitosterol

1 Products
Synonym:   α-Dihydrofucosterol, β-Sitosterol, 22,23-Dihydrostigmasterol, 24α-Ethylcholesterol, 5-Stigmasten-3β-ol
Image of Beta-sitosterol
CAS Number:
Empirical Formula (Hill Notation):
 C29H50O
Molecular Weight:
 414.71
 Beilstein/REAXYS Number:
 1916165
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More