• USA Home
  • Search Results

  • betaxololhydrochloride343896365919811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Betaxolol hydrochloride

4 Products
Synonym:   SL 75212
Image of Betaxolol hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C18H29NO3 · HCl
Molecular Weight:
 343.89
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Physical Form
  Special Grade
Show More