• USA Home
  • Search Results

  • betulin4427247398311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Betulin

3 Products
Synonym:   Betulinic alcohol, Betulinol, Betulol, Lup-20(29)-ene-3β,28-diol, Trochol
Image of Betulin
CAS Number:
Empirical Formula (Hill Notation):
 C30H50O2
Molecular Weight:
 442.72
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More