• USA Home
  • Search Results

  • betulinicacid4567047215111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Betulinic acid

4 Products
Synonym:   3β-Hydroxy-20(29)-lupaene-28-oic acid, Lupatic acid, Mairin
Image of Betulinic acid
CAS Number:
Empirical Formula (Hill Notation):
 C30H48O3
Molecular Weight:
 456.70
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Application(s)
Show More