• USA Home
  • Search Results

  • bglucuronidasefromhelixpomatia12345900145011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

β-Glucuronidase from Helix pomatia

7 Products
Synonym:   β-D-Glucuronide glucuronosohydrolase
Image of β-Glucuronidase from <I>Helix pomatia</I>
CAS Number:
 Enzyme Commission (EC) Number:
 3.2.1.31  ( BRENDA | IUBMB )
 MDL number:
 NACRES:
 NA.54
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More