• USA Home
  • Search Results

  • bglycerolphosphatedisodiumsaltpentahydrate306111340809811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

β-Glycerol phosphate disodium salt pentahydrate

1 Products
Synonym:   β-GP, β-glycerolphosphate, Disodium β-glycerol phosphate pentahydrate, glycerol-2-phosphate
Image of β-Glycerol phosphate disodium salt pentahydrate
CAS Number:
Empirical Formula (Hill Notation):
 C3H7Na2O6P · 5H2O
Molecular Weight:
 306.11
 Beilstein/REAXYS Number:
 3744328
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More