• USA Home
  • Search Results

  • bicalutamidecdx430379035706511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bicalutamide (CDX)

1 Products
Synonym:   Casodex, Cosudex, N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)sulfonyl]-2-hydroxy-2-methylpropanamide
Image of Bicalutamide (CDX)
CAS Number:
Empirical Formula (Hill Notation):
 C18H14F4N2O4S
Molecular Weight:
 430.37
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
Show More