• USA Home
  • Search Results

  • bicine1631715025411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bicine

3 Products
Synonym:   N,N-Bis(2-hydroxyethyl)glycine, BICINE
Image of Bicine
CAS Number:
Empirical Formula (Hill Notation):
 C6H13NO4
Molecular Weight:
 163.17
 Beilstein/REAXYS Number:
 1769362
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Absorption
  pH Value
  UV-Absorption Values
  Solubility
  Product Line
Show More