• USA Home
  • Search Results

  • bicyclo331nonane26dione152191647311311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bicyclo[3.3.1]nonane-2,6-dione

1 Products
Image of Bicyclo[3.3.1]<WBR>nonane-<WBR>2,6-<WBR>dione
CAS Number:
Empirical Formula (Hill Notation):
 C9H12O2
Molecular Weight:
 152.19
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More