• USA Home
  • Search Results

  • bifenazate3003514987741811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bifenazate

2 Products
Synonym:   N′-(4-Methoxybiphenyl-3-yl)hydrazinecarboxylic acid isopropyl ester
Image of Bifenazate
CAS Number:
Empirical Formula (Hill Notation):
 C17H20N2O3
Molecular Weight:
 300.35
 Beilstein/REAXYS Number:
 9632665
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Shelf Life
  Special Grade
Show More