• USA Home
  • Search Results

  • bifenthrin422879926718211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bifenthrin

2 Products
Synonym:   3-[(1Z)-2-Chloro-3,3,3-trifluoro-1-propenyl]-2,2-dimethylcyclopropanecarboxylic acid (2-methylbiphenyl-3-yl)methyl ester
Image of Bifenthrin
CAS Number:
Empirical Formula (Hill Notation):
 C23H22ClF3O2
Molecular Weight:
 422.87
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Shelf Life
  Special Grade
Show More