• USA Home
  • Search Results

  • bilirubin5846663565411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bilirubin

1 Products
Image of Bilirubin
CAS Number:
Empirical Formula (Hill Notation):
 C33H36N4O6
Molecular Weight:
 584.66
 Beilstein/REAXYS Number:
 74376
 EC Number:
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Physical Form
  Assay
Show More