• USA Home
  • Search Results

  • bionone192301490107611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

β-Ionone

4 Products
Synonym:   4-(2,6,6-Trimethyl-1-cyclohexenyl)-3-buten-2-one, beta-Ionone
Image of β-Ionone
CAS Number:
Empirical Formula (Hill Notation):
 C13H20O
Molecular Weight:
 192.30
 Beilstein/REAXYS Number:
 1909544
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More