• USA Home
  • Search Results

  • biotinphosphoramidite87807121750022511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Biotin Phosphoramidite

3 Products
Synonym:   Biotin amidite
Image of Biotin Phosphoramidite
CAS Number:
Empirical Formula (Hill Notation):
 C46H64N5O8PS
Molecular Weight:
 878.07
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Package quantity
  Product Line
  Compatibility
Show More