• USA Home
  • Search Results

  • biphenyl154219252411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Biphenyl

8 Products
Image of Biphenyl
CAS Number:
Linear Formula:
 C6H5C6H5
Molecular Weight:
 154.21
 Beilstein/REAXYS Number:
 1634058
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Agency/Method
  Shelf Life
  Package quantity
  Anal. Standard Form
  Product Line
  Assay
  Application(s)
  Physical Form
Show More