• USA Home
  • Search Results

  • bis24dinitrophenyloxalate422221653630411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bis(2,4-dinitrophenyl) oxalate

1 Products
Synonym:   2,4-Dinitrophenyl oxalate, DNPO, Oxalic acid bis(2,4-dinitrophenyl) ester
Image of Bis(2,4-<WBR>dinitrophenyl) oxalate
CAS Number:
Linear Formula:
 [-CO2C6H3(NO2)2]2
Molecular Weight:
 422.22
 Beilstein/REAXYS Number:
 2318321
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More