• USA Home
  • Search Results

  • bis2ethylhexyladipate3705710323111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Bis(2-ethylhexyl) adipate

6 Products
Synonym:   Adipic acid di(2-ethylhexyl) ester, DOA
Image of Bis(2-ethylhexyl) adipate
CAS Number:
Linear Formula:
 [-CH2CH2CO2CH2CH(C2H5)(CH2)3CH3]2
Molecular Weight:
 370.57
 Beilstein/REAXYS Number:
 1803774
 EC Number:
 MDL number:
 NACRES:
 NB.61
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Application(s)
  Potency
Show More